Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIGCHAP, MP Biomedicals™

Catalog No. ICN15229001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15229001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15229001 Supplier MP Biomedicals, Inc Supplier No. 0215229001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86303-22-2
Molecular Formula C42H75N3O16
Molecular Weight (g/mol) 878.067
InChI Key ZWEVPYNPHSPIFU-MNSMTYRASA-N
Synonym n,n'-bis 3-d-glucon-amidopropyl cholamide, n n-bis 3-d-gluconamido-propyl, bigchap n,n-bis 3-d-gluconamidopropyl-cholamide, 3a,5b,7a,12a-n,n-bis 3-d-gluconoylamino propyl-3,7,12-trihydroxycholan-24-amide
PubChem CID 122130704
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxy-N-[3-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl-[(4R)-4-[(3R,5S,7R,12S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentan
SMILES CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Quantity 1 g
Formula Weight 878.1
Physical Form Powder
Chemical Name or Material BIGCHAP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.