Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CHAP, Big, Calbiochem™,

Catalog No. 2009651GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-096-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-096-51GM Supplier MilliporeSigma™ Supplier No. 2009651GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86303-22-2 , 68-12-2
Molecular Formula C42H75N3O16
Molecular Weight (g/mol) 878.067
InChI Key ZWEVPYNPHSPIFU-MNSMTYRASA-N
Synonym N,N-bis-(3-D-Gluconamidopropyl)cholamide, CHAP, Big
PubChem CID 122130704
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxy-N-[3-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl-[(4R)-4-[(3R,5S,7R,12S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentan
SMILES CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥95% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.