CAS RN 86347-15-1
IUPAC Name:
hydrogen 5-[1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride
Synonyms:
medetomidine
Molecular Weight (g/mol):
236.74
Molecular Formula:
C13H17ClN2
InChi Key:
VPNGEIHDPSLNMU-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C1=CN=CN1)C1=CC=CC(C)=C1C
1
–
1
of
1
results
1
R&D Systems™ Medetomidine hydrochloride
CAS: 86347-15-1 Molecular Formula: C13H17ClN2