accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864249-60-5

IUPAC Name: 2-[(2-amino-3-phenylpropyl)amino]-3-methyl-5-(naphthalen-2-yl)-6-(pyridin-4-yl)-3,4-dihydropyrimidin-4-one
Synonyms: 2-[(2-amino-3-phenylpropyl)amino]-3-methyl-5-(naphthalen-2-yl)-6-(pyridin-4-yl)pyrimidin-4-one
Molecular Weight (g/mol): 461.57
Molecular Formula: C29H27N5O
InChi Key: RQVKVJIRFKVPBF-UHFFFAOYNA-N
SMILES: CN1C(NCC(N)CC2=CC=CC=C2)=NC(C2=CC=NC=C2)=C(C2=CC=C3C=CC=CC3=C2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results