CAS RN 86445-22-9
CAS RN 86445-22-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Auxinole
Auxin antagonist of TIR1/AFB receptors; also OsTIR1 inhibitor in auxin-inducible degron (AID) system