accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONBrBr

CAS RN 865163-47-9

IUPAC Name: 3,6-dibromo-9-[4-(dodecyloxy)phenyl]-9H-carbazole
Synonyms: 3,6-dibromo-9-[4-(dodecyloxy)phenyl]carbazole
Molecular Weight (g/mol): 585.42
Molecular Formula: C30H35Br2NO
InChi Key: VWCTWVHXOOAKSE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results