CAS RN 866396-70-5
IUPAC Name:
1-(2-{[1-(hydroxymethyl)cyclopentyl]amino}acetyl)pyrrolidine-2,5-dicarbonitrile hydrochloride
Synonyms:
1-(2-{[1-(hydroxymethyl)cyclopentyl]amino}acetyl)pyrrolidine-2,5-dicarbonitrile hydrochloride
Molecular Weight (g/mol):
312.8
Molecular Formula:
C14H21ClN4O2
InChi Key:
RHCWAHDCHQTNBR-UHFFFAOYNA-N
SMILES:
Cl.OCC1(CCCC1)NCC(=O)N1C(CCC1C#N)C#N
1
–
1
of
1
results
1
R&D Systems™ DPPI 1c hydrochloride
CAS: 866396-70-5 Molecular Formula: C14H21ClN4O2