CAS RN 866914-87-6
IUPAC Name:
N'-[4-(2-{[(3-chlorophenyl)methyl]amino}ethyl)phenyl]thiophene-2-carboximidamide dihydrochloride
Synonyms:
N'-[4-(2-{[(3-chlorophenyl)methyl]amino}ethyl)phenyl]thiophene-2-carboximidamide dihydrochloride
Molecular Weight (g/mol):
442.83
Molecular Formula:
C20H22Cl3N3S
InChi Key:
SOXBYIKSUDXMIE-UHFFFAOYSA-N
SMILES:
Cl.Cl.NC(=NC1=CC=C(CCNCC2=CC(Cl)=CC=C2)C=C1)C1=CC=CS1
1
–
1
of
1
results
1
R&D Systems™ ARL 17477 dihydrochloride
CAS: 866914-87-6 Molecular Formula: C20H22Cl3N3S