accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSH

CAS RN 86704-82-7

IUPAC Name: 3-ethoxybenzene-1-thiol
Synonyms: 3-ethoxybenzenethiol
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: RTJMJGJLVSKSEB-UHFFFAOYSA-N
SMILES: CCOC1=CC(S)=CC=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)