CAS RN 867160-71-2
CAS RN 867160-71-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Linsitinib

Potent and selective dual inhibitor of the IGF-1 receptor and insulin receptor