accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOH3C

CAS RN 868-85-9

IUPAC Name: dimethoxy(oxo)-λ⁵-phosphanylium
Synonyms: dimethoxy(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 109.04
Molecular Formula: C2H6O3P
InChi Key: YLFBFPXKTIQSSY-UHFFFAOYSA-N
SMILES: CO[P+](=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)