CAS RN 868265-28-5
IUPAC Name:
2-({2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butylphenoxy]ethyl}(methyl)amino)acetic acid hydrochloride
Synonyms:
({2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butylphenoxy]ethyl}(methyl)amino)acetic acid hydrochloride
Molecular Weight (g/mol):
421.92
Molecular Formula:
C22H28ClNO5
InChi Key:
ZQVOAGQZHDAFRM-UHFFFAOYSA-N
SMILES:
Cl.CN(CCOC1=C(C=C(C=C1)C1=CC2=C(OCO2)C=C1)C(C)(C)C)CC(O)=O
1
–
1
of
1
results
1
R&D Systems™ LY 2365109 hydrochloride
CAS: 868265-28-5 Molecular Formula: C22H28ClNO5