CAS RN 868540-17-4
IUPAC Name:
4-methyl-N-(1-{[4-methyl-1-(2-methyloxiran-2-yl)-1-oxopentan-2-yl]carbamoyl}-2-phenylethyl)-2-{2-[2-(morpholin-4-yl)acetamido]-4-phenylbutanamido}pentanamide
Synonyms:
carfilzomib
Molecular Weight (g/mol):
719.92
Molecular Formula:
C40H57N5O7
InChi Key:
BLMPQMFVWMYDKT-UHFFFAOYNA-N
SMILES:
CC(C)CC(NC(=O)C(CCC1=CC=CC=C1)NC(=O)CN1CCOCC1)C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC(C)C)C(=O)C1(C)CO1
1
–
2
of
2
results
1
R&D Systems™ Carfilzomib
A Tocris product, part of R&D Systems. Potent irreversible proteasome inhibitor; active in vivo
2
Promotions Available
Carfilzomib, Thermo Scientific™
Carfilzomib, Thermo Scientific™
CAS: 868540-17-4 Molecular Formula: C40 H57 N5 O7