accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3C

CAS RN 86917-58-0

IUPAC Name: 1,5-dimethylpiperidin-2-one
Synonyms: 1,5-dimethylpiperidin-2-one
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: YXHVIDNQBMVYHQ-UHFFFAOYNA-N
SMILES: CC1CCC(=O)N(C)C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results