accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOHO

CAS RN 87-20-7

IUPAC Name: 3-methylbutyl 2-hydroxybenzoate
Synonyms: isoamyl salicylate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: PMGCQNGBLMMXEW-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results