accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFFONSN

CAS RN 87051-43-2

IUPAC Name: 6-(2-{4-[bis(4-fluorophenyl)methylidene]piperidin-1-yl}ethyl)-7-methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
Synonyms: ritanserin ritanserin tartrate tiserton 利坦色林 ritanserinum ritanserine ritanserina
Molecular Weight (g/mol): 477.57
Molecular Formula: C27H25F2N3OS
InChi Key: JUQLTPCYUFPYKE-UHFFFAOYSA-N
SMILES: CC1=C(CCN2CCC(CC2)=C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(=O)N2C=CSC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)