accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrFOH2N

CAS RN 871353-25-2

IUPAC Name: 3-bromo-2-fluorobenzamide
Synonyms: 3-bromo-2-fluorobenzamide
Molecular Weight (g/mol): 218.03
Molecular Formula: C7H5BrFNO
InChi Key: UWTBFBQQCCZYBF-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results