accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNOCH3H3C

CAS RN 871839-91-7

IUPAC Name: 2-(propan-2-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 263.14
Molecular Formula: C14H22BNO3
InChi Key: WCAIJTJWLUYXIW-UHFFFAOYSA-N
SMILES: CC(C)OC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results