accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrOH(E/Z)(E/Z)

CAS RN 872201-12-2

IUPAC Name: 4-(2-{3-bromo-4-[2-(4-hydroxyphenyl)ethenyl]phenyl}ethenyl)phenol
Synonyms: 4-(2-{3-bromo-4-[2-(4-hydroxyphenyl)ethenyl]phenyl}ethenyl)phenol
Molecular Weight (g/mol): 393.28
Molecular Formula: C22H17BrO2
InChi Key: OXPHQQMZTXMEGO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=CC2=CC(Br)=C(C=CC3=CC=C(O)C=C3)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results