CAS RN 87227-41-6
IUPAC Name:
2-(4-bromobutyl)-1,3-dioxolane
Synonyms:
2-(4-bromobutyl)-1,3-dioxolane
Molecular Weight (g/mol):
209.08
Molecular Formula:
C7H13BrO2
InChi Key:
UQTZEBQZCBQLAN-UHFFFAOYSA-N
SMILES:
BrCCCCC1OCCO1
1
–
1
of
1
results
1
2-(4-Bromobutyl)-1,3-dioxolane 95.0+%, TCI America™
CAS: 87227-41-6 Molecular Formula: C7H13BrO2 Molecular Weight (g/mol): 209.083 MDL Number: MFCD02093454 InChI Key: UQTZEBQZCBQLAN-UHFFFAOYSA-N Synonym: 5-Bromovaleraldehyde Ethylene Acetal PubChem CID: 13780200 IUPAC Name: 2-(4-bromobutyl)-1,3-dioxolane SMILES: C1COC(O1)CCCCBr