Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromobutyl)-1,3-dioxolane 95.0+%, TCI America™
SDP

Catalog No. B21285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21285G 5 g
B212825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21285G Supplier TCI America Supplier No. B21285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87227-41-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD02093454
InChI Key UQTZEBQZCBQLAN-UHFFFAOYSA-N
Synonym 5-Bromovaleraldehyde Ethylene Acetal
PubChem CID 13780200
IUPAC Name 2-(4-bromobutyl)-1,3-dioxolane
SMILES C1COC(O1)CCCCBr

Specifications

Color Yellow
Quantity 5 g
Formula Weight 209.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Bromobutyl)-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.