accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87239-81-4

IUPAC Name: 1-{[(propan-2-yloxy)carbonyl]oxy}ethyl (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cefpodoxime proxetil
Molecular Weight (g/mol): 557.59
Molecular Formula: C21H27N5O9S2
InChi Key: LTINZAODLRIQIX-FBXRGJNPSA-N
SMILES: COCC1=C(N2[C@H](SC1)[C@H](NC(=O)C(=N/OC)\C1=CSC(N)=N1)C2=O)C(=O)OC(C)OC(=O)OC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)