Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, 25% aq. soln.

Catalog No. AAJ6606103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6606103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6606103 Supplier Thermo Scientific Chemicals Supplier No. J6606103
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 87239-81-4 | C21H27N5O9S2 | 557.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87239-81-4
Molecular Formula C21H27N5O9S2
Molecular Weight (g/mol) 557.593
MDL Number MFCD00865088
InChI Key LTINZAODLRIQIX-FBXRGJNPSA-N
PubChem CID 6526396
ChEBI CHEBI:3505
IUPAC Name 1-propan-2-yloxycarbonyloxyethyl (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(C)OC(=O)OC(C)OC(=O)C1=C(CSC2N1C(=O)C2NC(=O)C(=NOC)C3=CSC(=N3)N)COC

Specifications

Type Cefpodoxime Proxetil
Physical Form Powder
Melting Point 111°C to 113°C
Color White
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in DMSO
Formula Weight 557.6
Percent Purity 96%
Chemical Name or Material Cefpodoxime proxetil
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
RTECSNumber XI0367370
TSCA No
storageNote1 2° to 8°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.