Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefpodoxime Proxetil, G-Biosciences
SCI_ED

Catalog No. S963101
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S963101 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S963101 Supplier G-Biosciences Supplier No. RC938
Add to Cart
Add to Cart

Cefpodoxime proxetil is a third generation cephalosporin antibiotic targeting a wide variety of gram-negative and gram-positive organisms, especially those responsible for otitis media and pharyngitis.

It is an ester prodrug of the free acid metabolite cefpodoxime.

Chemical Identifiers

CAS 87239-81-4
Molecular Weight (g/mol) 557.60 g/mol

Specifications

Technique HPLC, UV/Vis, Spectrometry, Ion Chromatography, Gas Chromatography
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 100 mg
Percent Purity >95%
Grade Molecular Biology
Chemical Name or Material Cefpodoxime Proxetil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.