accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874-18-0

IUPAC Name: 2,4-dibromo-6-chloroaniline
Synonyms: 2,4-dibromo-6-chloroaniline
Molecular Weight (g/mol): 285.36
Molecular Formula: C6H4Br2ClN
InChi Key: MFROLNRQDCAZCD-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Br)C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results