CAS RN 874-23-7
CAS RN 874-23-7
2-Acetylcyclohexanone, 97%
CAS: 874-23-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00001633 InChI Key: OEKATORRSPXJHE-UHFFFAOYNA-N Synonym: 2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone PubChem CID: 13400 IUPAC Name: 2-acetylcyclohexan-1-one SMILES: CC(=O)C1CCCCC1=O
2-Acetylcyclohexanone 97.0+%, TCI America™
CAS: 874-23-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00001633 InChI Key: OEKATORRSPXJHE-UHFFFAOYNA-N Synonym: 2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone PubChem CID: 13400 IUPAC Name: 2-acetylcyclohexan-1-one SMILES: CC(=O)C1CCCCC1=O
Sigma Aldrich 2-Acetylcyclohexanone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2-Acetylcyclohexanone, 98%, Thermo Scientific™
CAS: 874-23-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00001633 InChI Key: OEKATORRSPXJHE-UHFFFAOYNA-N PubChem CID: 13400 IUPAC Name: 2-acetylcyclohexan-1-one SMILES: CC(=O)C1CCCCC1=O
2-Acetylcyclohexanone, 98%, Thermo Scientific™
CAS: 874-23-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00001633 InChI Key: OEKATORRSPXJHE-UHFFFAOYNA-N Synonym: 2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone PubChem CID: 13400 IUPAC Name: 2-acetylcyclohexan-1-one SMILES: CC(=O)C1CCCCC1=O