Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylcyclohexanone, 98%, Thermo Scientific™

Catalog No. AAL0275306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0275306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0275306 Supplier Thermo Scientific Chemicals Supplier No. L0275306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Acetylcyclohexanone is useful for synthesis of anilinoethanolamines, 1-(pyridyl)ethanol derivatives that can reduce keto-esters and ketones. It is also used in the synthesis of phenothiazine derivatives that can inhibit prote in farnesyltransferase molecules and some that show anti-proliferative activity.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, Strong bases. Air-sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-23-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001633
InChI Key OEKATORRSPXJHE-UHFFFAOYNA-N
PubChem CID 13400
IUPAC Name 2-acetylcyclohexan-1-one
SMILES CC(=O)C1CCCCC1=O

Specifications

Density 1.063
Boiling Point 111°C to 112°C (18 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.511
Quantity 5 g
Beilstein 1858621
Synonym 2-acetylcyclohexanone, cyclohexanone, 2-acetyl, 2-acetyl-1-cyclohexanone, .alpha.-acetylcyclohexanone, 2-acetyl cyclohexanone, 2-acetyl-cyclohexanone, cyclohexanone,2-acetyl, 1-acetyl-2-oxocyclohexane, alpha-acetylcyclohexanone, acetylcyclohexanone
Solubility Information Not miscible in water.
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material 2-Acetylcyclohexanone
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 212-858-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.