Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylcyclohexanone, 98%, Thermo Scientific™

Catalog No. AAA1170106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1170106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1170106 Supplier Thermo Scientific Chemicals Supplier No. A1170106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-23-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001633
InChI Key OEKATORRSPXJHE-UHFFFAOYNA-N
Synonym 2-acetylcyclohexanone, cyclohexanone, 2-acetyl, 2-acetyl-1-cyclohexanone, .alpha.-acetylcyclohexanone, 2-acetyl cyclohexanone, 2-acetyl-cyclohexanone, cyclohexanone,2-acetyl, 1-acetyl-2-oxocyclohexane, alpha-acetylcyclohexanone, acetylcyclohexanone
PubChem CID 13400
IUPAC Name 2-acetylcyclohexan-1-one
SMILES CC(=O)C1CCCCC1=O

Specifications

Boiling Point 111°C to 112°C (18 mmHg)
Flash Point 79°C (175°F)
Quantity 5 g
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material 2-Acetylcyclohexanone
Hazard Category Warning
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.