CAS RN 874-33-9
IUPAC Name:
methyl[(2-methylphenyl)methyl]azanium
Synonyms:
methyl[(2-methylphenyl)methyl]azanium
Molecular Weight (g/mol):
136.22
Molecular Formula:
C9H14N
InChi Key:
YMWQUYQBTXWNAH-UHFFFAOYSA-O
SMILES:
C[NH2+]CC1=CC=CC=C1C
1
–
1
of
1
results
1
N-Methyl-2-methylbenzylamine, 95%, Thermo Scientific™
CAS: 874-33-9 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 MDL Number: MFCD04625428 InChI Key: YMWQUYQBTXWNAH-UHFFFAOYSA-N Synonym: n-methyl-1-o-tolyl methanamine,n-methyl-n-2-methylbenzyl amine,2-methyl-n-methylbenzylamine,n-methyl-1-2-methylphenyl methanamine,n,2-dimethylbenzylamine,methyl 2-methylphenyl methyl amine,benzenemethanamine, n,2-dimethyl,methyl 2-methylbenzyl amine,ksc493q4r PubChem CID: 263103 IUPAC Name: N-methyl-1-(2-methylphenyl)methanamine SMILES: CC1=CC=CC=C1CNC