Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-2-methylbenzylamine, 95%, Thermo Scientific™

Catalog No. AAH6395603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6395603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6395603 Supplier Thermo Scientific Chemicals Supplier No. H6395603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-N-methyl-2-methylbenzylamine is resistant to displacement by the aqueous extractants.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-33-9
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD04625428
InChI Key YMWQUYQBTXWNAH-UHFFFAOYSA-N
Synonym n-methyl-1-o-tolyl methanamine, n-methyl-n-2-methylbenzyl amine, 2-methyl-n-methylbenzylamine, n-methyl-1-2-methylphenyl methanamine, n,2-dimethylbenzylamine, methyl 2-methylphenyl methyl amine, benzenemethanamine, n,2-dimethyl, methyl 2-methylbenzyl amine, ksc493q4r
PubChem CID 263103
IUPAC Name N-methyl-1-(2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CNC

Specifications

Quantity 1 g
UN Number UN2735
Solubility Information Slightly soluble in water.
Formula Weight 135.21
Percent Purity 95%
Chemical Name or Material N-Methyl-2-methylbenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.