accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874-97-5

IUPAC Name: 3-(hydroxymethyl)benzonitrile
Synonyms: 3-(hydroxymethyl)benzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: PCOFIIVWHXIDGT-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C#N
Percent Purity 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results