CAS RN 874882-72-1
IUPAC Name:
4-[3-(tert-butylamino)-2-hydroxypropyl]-1H-indole-2-carbonitrile; but-2-enedioic acid
Synonyms:
4-[3-(tert-butylamino)-2-hydroxypropyl]-1H-indole-2-carbonitrile; butenedioic acid
Molecular Weight (g/mol):
387.44
Molecular Formula:
C20H25N3O5
InChi Key:
IOZROHWNOAFEMY-UHFFFAOYNA-N
SMILES:
OC(=O)C=CC(O)=O.CC(C)(C)NCC(O)CC1=C2C=C(NC2=CC=C1)C#N
1
–
1
of
1
results
1
R&D Systems™ Cyanopindolol hemifumarate
CAS: 874882-72-1 Molecular Formula: C20H25N3O5