CAS RN 874882-93-6
IUPAC Name:
4-[5-(4-chlorophenyl)-4-methyl-1,2-oxazol-3-yl]-1-(2-phenylethyl)piperidine hydrochloride
Synonyms:
4-[5-(4-chlorophenyl)-4-methyl-1,2-oxazol-3-yl]-1-(2-phenylethyl)piperidine hydrochloride
Molecular Weight (g/mol):
417.37
Molecular Formula:
C23H26Cl2N2O
InChi Key:
HZRPUQURUAXOHB-UHFFFAOYSA-N
SMILES:
Cl.CC1=C(ON=C1C1CCN(CCC2=CC=CC=C2)CC1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ L-741,742 hydrochloride
CAS: 874882-93-6 Molecular Formula: C23H26Cl2N2O