accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNClN

CAS RN 875-35-4

IUPAC Name: 2,6-dichloro-4-methylpyridine-3-carbonitrile
Synonyms: 2,6-dichloro-4-methylpyridine-3-carbonitrile
Molecular Weight (g/mol): 187.02
Molecular Formula: C7H4Cl2N2
InChi Key: LSPMHHJCDSFAAY-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC(Cl)=C1C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)