accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876317-18-9

IUPAC Name: 1-[4-(thiophen-3-yl)phenyl]methanamine
Synonyms: 1-[4-(thiophen-3-yl)phenyl]methanamine
Molecular Weight (g/mol): 189.28
Molecular Formula: C11H11NS
InChi Key: QTXFTOUAVPVJMZ-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C1)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results