accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOONFFFFFFF

CAS RN 877052-79-4

IUPAC Name: 4-{[3,5-bis(trifluoromethyl)phenyl]methyl}-6-(2-fluorophenyl)-2H,3H,4H,5H-pyrido[3,2-f][1,4]oxazepin-3-one
Synonyms: 4-{[3,5-bis(trifluoromethyl)phenyl]methyl}-6-(2-fluorophenyl)-2H,5H-pyrido[3,2-f][1,4]oxazepin-3-one
Molecular Weight (g/mol): 484.37
Molecular Formula: C23H15F7N2O2
InChi Key: ZIXNJVGTAXRKAP-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=C2CN(CC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(=O)COC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results