accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNSOOOONOO

CAS RN 877636-42-5

IUPAC Name: 4-oxo-6-[(pyrimidin-2-ylsulfanyl)methyl]-4H-pyran-3-yl 4-nitrobenzoate
Synonyms: 4-oxo-6-[(pyrimidin-2-ylsulfanyl)methyl]pyran-3-yl 4-nitrobenzoate
Molecular Weight (g/mol): 385.35
Molecular Formula: C17H11N3O6S
InChi Key: UASIRTUMPRQVFY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)OC1=COC(CSC2=NC=CC=N2)=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results