accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 878483-02-4

IUPAC Name: tert-butyl 3-({7-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl}oxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: tert-butyl 3-({7-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl}oxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 658.71
Molecular Formula: C35H38N4O9
InChi Key: FJYOABRVKHDUNV-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(COC1OC2COC(OC2C(O)C1N=[N+]=[N-])C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results