accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHNHSOOCH3OCl

CAS RN 878489-28-2

IUPAC Name: 2-(4-chlorophenoxy)-N-(5-methanesulfonamidoadamantan-2-yl)-2-methylpropanamide
Synonyms: 2-(4-chlorophenoxy)-N-(5-methanesulfonamidoadamantan-2-yl)-2-methylpropanamide
Molecular Weight (g/mol): 440.98
Molecular Formula: C21H29ClN2O4S
InChi Key: OSGIRCJRKSAODN-UHFFFAOYNA-N
SMILES: CC(C)(OC1=CC=C(Cl)C=C1)C(=O)NC1C2CC3CC1CC(C3)(C2)NS(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results