accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 878745-51-8

IUPAC Name: methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
Synonyms: methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
Molecular Weight (g/mol): 206.12
Molecular Formula: C7H5F3N2O2
InChi Key: VBFLWQBZJODKRL-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(=NC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results