accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFMgBr

CAS RN 879-05-0

IUPAC Name: bromo(2,3,4,5,6-pentafluorophenyl)magnesium
Synonyms: bromo(2,3,4,5,6-pentafluorophenyl)magnesium
Molecular Weight (g/mol): 271.27
Molecular Formula: C6BrF5Mg
InChi Key: AXQVMJIDNDRSFM-UHFFFAOYSA-M
SMILES: FC1=C(F)C(F)=C([Mg]Br)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results