Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenylmagnesium bromide, 0.5M solution in diethyl ether, AcroSeal™

Catalog No. AC438741000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438741000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438741000 Supplier Thermo Scientific Chemicals Supplier No. 438741000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 879-05-0 | C6BrF5Mg | 271.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879-05-0 , 60-29-7
Molecular Formula C6BrF5Mg
Molecular Weight (g/mol) 271.27
MDL Number MFCD00015720
InChI Key AXQVMJIDNDRSFM-UHFFFAOYSA-M
Synonym pentafluorophenylmagnesium bromide, magnesium, bromo pentafluorophenyl, pentafluorophenylmagnesiumbromide, c6f5mgbr, pentafluorophenyl magnesiumbromide, axqvmjidndrsfm-uhfffaoysa-m, pentafluorophenyl magnesium bromide, pentafluorophenylmagnesium bromide solution, bromo 2,3,4,5,6-pentafluorophenyl magnesium, pentafluorophenylmagnesium bromide solution, 0.5 m in diethyl ether, grignard reagent
PubChem CID 3319152
IUPAC Name magnesium;1,2,3,4,5-pentafluorobenzene-6-ide;bromide
SMILES FC1=C(F)C(F)=C([Mg]Br)C(F)=C1F

Specifications

Density 0.8600g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula F5C6MgBr
Quantity 100 mL
Specific Gravity 0.86
Formula Weight 271.27
Chemical Name or Material Pentafluorophenylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes,on s

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.