CAS RN 879089-64-2
IUPAC Name:
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H,4H,5H-pyrrolo[3,2-b]pyridin-5-one hydrate dihydrochloride
Synonyms:
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H,4H-pyrrolo[3,2-b]pyridin-5-one hydrate dihydrochloride
Molecular Weight (g/mol):
306.19
Molecular Formula:
C12H17Cl2N3O2
InChi Key:
UXDAADMPPAHNMY-UHFFFAOYSA-N
SMILES:
O.Cl.Cl.O=C1NC2=C(NC=C2C2=CCNCC2)C=C1
1
–
1
of
1
results
1
R&D Systems™ CP 93129 dihydrochloride
CAS: 879089-64-2 Molecular Formula: C12H17Cl2N3O2