accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClSHOOO

CAS RN 88-43-7

IUPAC Name: 5-amino-2-chlorobenzene-1-sulfonic acid
Synonyms: 5-amino-2-chlorobenzenesulfonic acid
Molecular Weight (g/mol): 207.63
Molecular Formula: C6H6ClNO3S
InChi Key: VPXCXBHLKDPWQV-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results