Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroaniline-3-sulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. C0116500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0116500G 500 g
C011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0116500G Supplier TCI America Supplier No. C0116500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-43-7
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.628
MDL Number MFCD00025296
InChI Key VPXCXBHLKDPWQV-UHFFFAOYSA-N
Synonym 5-Amino-2-chlorobenzenesulfonic Acid
PubChem CID 6933
IUPAC Name 5-amino-2-chlorobenzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)Cl

Specifications

Quantity 500 g
Formula Weight 207.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloroaniline-3-sulfonic Acid
EINECSNumber (3)-2019
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.