accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHONOONOO

CAS RN 88-85-7

IUPAC Name: 2-(butan-2-yl)-4,6-dinitrophenol
Synonyms: caldon dinoseb
Molecular Weight (g/mol): 240.22
Molecular Formula: C10H12N2O5
InChi Key: OWZPCEFYPSAJFR-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
Concentration 
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)