Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinoseb, Restek
SDP

Catalog No. 06719876 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-876 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-876 Supplier Restek Corporation Supplier No. 32251
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-85-7
Molecular Formula C10H12N2O5
Molecular Weight (g/mol) 240.215
InChI Key OWZPCEFYPSAJFR-UHFFFAOYSA-N
PubChem CID 6950
ChEBI CHEBI:83632
IUPAC Name 2-butan-2-yl-4,6-dinitrophenol
SMILES CCC(C)C1=CC(=CC(=C1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Packaging Ampule
Concentration 1000 μg/mL
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material Dinoseb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.