Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinoseb Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USHB1301
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HB-130-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HB-130-1 Supplier Agilent Technologies Supplier No. HB1301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-85-7 , 67-56-1
Molecular Formula C10H12N2O5
Molecular Weight (g/mol) 240.215
InChI Key OWZPCEFYPSAJFR-UHFFFAOYSA-N
PubChem CID 6950
ChEBI CHEBI:83632
IUPAC Name 2-butan-2-yl-4,6-dinitrophenol
SMILES CCC(C)C1=CC(=CC(=C1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Max % 99.98, 0.01
Analyte or Component Names Methanol,Dinoseb
Element Methanol,Dinoseb
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Dinoseb Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.