CAS RN 885060-09-3
IUPAC Name:
2-(3-aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-2,3-dihydro-1,3,4-thiadiazole-3-carboxamide
Synonyms:
filanesib
Molecular Weight (g/mol):
420.48
Molecular Formula:
C20H22F2N4O2S
InChi Key:
LLXISKGBWFTGEI-UHFFFAOYNA-N
SMILES:
CON(C)C(=O)N1N=C(SC1(CCCN)C1=CC=CC=C1)C1=C(F)C=CC(F)=C1
1
–
1
of
1
results
1
R&D Systems™ ARRY 520 trifluoroacetate
CAS: 885060-09-3 Molecular Formula: C20H22F2N4O2S