accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOCH3N

CAS RN 88569-83-9

IUPAC Name: 6-methoxy-N-methylpyridin-2-amine
Synonyms: 6-methoxy-N-methylpyridin-2-amine
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: GXHQLRBOZRJHPZ-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(OC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results